Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
- Fixative 1 vial/12mL Room Temperature
- SREBP-2 Primary Antibody 1 vial/100μL -20°C
- Blocking Solution 1 vial/12mL 4°C
- DyLightTM 549-Conjugated Goat Anti-Rabbit Secondary Antibody 1 vial/100μL -20°C
- U18666A, 2.5mM 1 vial/100μL -20°C
Specifications
Specifications
| Content And Storage | -20°C |
| For Use With (Application) | Immunofluorescence, functional study |
| Includes | 12mL Fixative, 100μL SREBP-2 primary antibody, 12mL blocking solution, 100μL DyLight 549 conjugated Goat anti-Rabbit secondary antibody, 100μL U18666A (2.5mM) |
| Quantity | 96 Assays |
| Type | Cell-Based Translocation Assay Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?