Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Sulfate Assay Kit

Catalog No. 89028353 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-353 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-353 Supplier Abnova™ Supplier No. KA1621
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Reagent A, 25mL
  • Reagent B, 2.4g powder
  • TCA Reagent, 25mL
  • Sulfate standard, 1mL (60mM)

For quantitative turbidimetric sulfate determination at 600nm in plasma, serum, urine

Specifications

Calibration Range 0.01 to 1.2mm
Content And Storage Store the kit at room temperature.
Description Sulfate Assay Kit is a quantitative turbidimetric sulfate determination at 600 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 25mL Reagent A, 2.4g powder reagent B, 25mL TCA reagent, 1mL 60mM Sulfate standard
Quantity 1 Kit
Sample Type Plasma, Serum, Urine
Sample Volume 600 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.