Learn More
Specifications
Specifications
| Content And Storage | Store the kit at -20°C. Store SP conjugate, Biotinylated antibody and Reconstituted Standard at -20°C, other components may be stored at 4°C. Avoid repeated freezing and thawing. |
| Description | TF (Human) Chromogenic Activity Assay Kit is developed to determine human TF chromogenic activity in plasma, tissue, and cell culture supernatants. |
| For Use With (Application) | Functional Study |
| Includes | 96-well Polystyrene microplate, sealing tapes, 30mL sample diluent, 30mL assay diluent, 1 vial rhTF standard (Lipoprotein), 1 vial human FVII, 1 vial human FX, 2 vials FXa substrate |
| Quantity | 1 Kit |
| Sample Type | Plasma, Tissue and Cell Culture Supernatants |
| Type | Activity Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.