Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Tf (Rat) ELISA Kit

Catalog No. 89028598 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
HRP
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
89-028-598 Biotin
89-100-703 HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-598 Supplier Abnova™ Supplier No. KA1847
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of rat Tf

Specifications

Conjugate Biotin
Cross Reactivity Rat
Description Tf (Rat) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of rat Tf.
Detection Limit 150 ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, rat TF standard 20μg, Biotinylated TF 1 vial, 30mL MIX diluent concentrate (10X), 30mL Wash Buffer 20X, Streptavidin peroxidase Conjugate (SP Conjugate) 80, 8mL Chromogen Substrate, 12mL stop solution
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 25 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type TF (Rat) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.