Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TGFA (Human) ELISA Kit

Catalog No. 89100808 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Body Fluid, CCS, Serum, Tissue Lysates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-808 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-027-206 Body Fluid, CCS, Serum, Tissue Lysates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-808 Supplier Abnova™ Supplier No. KA3105
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human TGFA

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description TGFA (Human) ELISA Kit is intended for the quantitative measurement of human TGFA.
Detection Limit 1pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Human TGF alpha antibody, Lyophilized TGF alpha standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Human TGF alpha antibody (Concentrated Dilution: 1:100) (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated Dilution: 1:100) (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type TGFA (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.