Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Thiol Detection Assay Kit

Catalog No. 89024502 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-502 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-502 Supplier Abnova™ Supplier No. KA1382
Add to Cart
Add to Cart

Includes

  • Thiol assay buffer (10X), 5mL
  • Thiol fluorometric detector, 100μL
  • Thiol GSH standard, 10mg
  • Thiol cysteine standard, 10mg
  • Half volume plate (Black), 96 wells
  • 96-Well cover sheet, 1 cover

Used for the detection of free thiol content at excitation wavelengths from 380 to 390nm and emmision wavelengths from 510 to 520nm

Thiol Detection Assay Kit provides a method for assaying free thiol content in plasma, serum, tissue homogenates, cell lysates, and urine.

Functional Study

Specifications

Content And Storage Store the kit at -20°C.
Description Thiol Detection Assay Kit is used for the detection of free thiol content at excitation wavelengths between 380-390 nm and emmision wavelengths between 510-520 nm.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 5mL Thiol assay buffer 10X, 100μL Thiol fluorometric detector, 10mg Thiol GSH standard, 10mg Thiol Cysteine standard, 96 wells half volume plate (black), 1 cover 96-well cover sheet
Quantity 1 Kit
Sample Type Plasma, Serum, Tissue Homogenates, Cell Lysates and Urine
Sample Volume 50 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.