Learn More
Abnova™ Total Histone H4 Acetylation Detection Fast Kit (Colorimetric)
Shop All Abnova Corporation ProductsDescription
- Quick and efficient procedure, which can be finished within 2.5 hours
- Innovative colorimetric assay without the need for radioactivity, electrophoresis, or chromatography
- Captures histone H4 acetylated at any lysine site with the detection limit as low as 2 ng/well and detection range from 5 ng-2 μg/well of histone extracts
- The control is conveniently included for the quantification of the amount of acetylated histone H4
- Strip microplate format makes the assay flexible: manual or high throughput
- Simple, reliable, and consistent assay conditions
- Upon receipt store C3 and Standard Control at -20°C; store all other components at 4°C away from light
- The kit is stable for up to 6 months from the shipment date, when stored properly
- For research use only
Specifications
Specifications
| Description | Total Histone H4 Acetylation Detection Fast Kit (Colorimetric) is used for measuring total acetylation of histone H4. |
| Product Type | Total Histone H4 Acetylation Detection Fast Kit (Colorimetric) |
| Sample Type | Histone Extract |
| Specificity | Histone extracts from mammalian cells/ tissues |
| For Use With (Application) | Functional Study |
| Quantity | 1 Kit |
| Sample Volume | 50 μL |
| Storage Requirements | Store the detection antibody and the standard at -20°C. Store all other components at 4°C away from light. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.