Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
TP53 Cell-Based Activation/Translocation Assay Kit provides a highly specific phospho-p53 MAPK primary monoclonal antibody together with a Dylight™ conjugated secondary antibody.
Immunofluorescence, Functional Study
Specifications
Specifications
| Content And Storage | -20°C |
| For Use With (Application) | Immunofluorescence, functional study |
| Includes | 1 vial fixative, 1 vial p53 total monoclonal primary antibody, 1 vial blocking solution, 1 vial DyLight 549 conjugated goat anti-mouse secondary antibody, 1 vial (-)-Nutlin-3 (10mM) |
| Quantity | 100 Tests |
| Type | Cell-Based Activation/Translocation Assay Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?