Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TSH (Human) ELISA Kit

Catalog No. 89100043 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Serum
Calibration Range:
0 to 10uIU/mL
0 to 25uIU/mL
0.05 to 12μIU/mL
0.2 to 40μIU/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type Calibration Range
89-100-043 Serum 0 to 10uIU/mL
89-100-044 Serum 0 to 25uIU/mL
89-100-209 Serum 0.05 to 12μIU/mL
89-100-211 Serum 0.2 to 40μIU/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 89-100-043 Supplier Abnova™ Supplier No. KA0197
Add to Cart
Add to Cart

Includes

  • Murine Monoclonal Anti-TSH coated Microtiter wells, 1 plate, 96 wells
  • Reference Standard Set: Contains 0, 0.1, 0.5, 2, 5, and 10μIU/mL, lyophilized
  • Enzyme Conjugate Reagent, 13mL
  • TMB Reagent (One-Step), 11mL
  • Stop Solution (1N HCl), 11mL

The ELISA Test is based on principle of a solid phase enzyme-linked immunosorbent assay

Specifications

Calibration Range 0 to 10uIU/mL
Conjugate HRP
Cross Reactivity Human
Description TSH (Human) High Sensitive ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of thyroid stimulating hormone.
Detection Limit 0.07uIU/mL
Detection Method Colorimetric
Includes Murine Monoclonal Anti-TSH coated Microtiter wells. 1 plate, 96 wells, Reference Standard Set: Contains 0, 0.1, 0.5, 2, 5, and 10μIU/ml. Lyophilized, Enzyme Conjugate Reagent (13mL), TMB Reagent (One-Step) (11mL), Stop Solution (1N HCl) (11mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.