Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TSH (Human) ELISA Kit

Catalog No. 89100044 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Serum
Calibration Range:
0 to 10uIU/mL
0 to 25uIU/mL
0.05 to 12μIU/mL
0.2 to 40μIU/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type Calibration Range
89-100-044 Serum 0 to 25uIU/mL
89-100-043 Serum 0 to 10uIU/mL
89-100-209 Serum 0.05 to 12μIU/mL
89-100-211 Serum 0.2 to 40μIU/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 89-100-044 Supplier Abnova™ Supplier No. KA0196
Add to Cart
Add to Cart

Includes

  • Microtiter wells coated with murine monoclonal anti-TSH, 96 wells
  • Enzyme Conjugate Reagent, 1 vial (13mL)
  • Reference Standard Set : 0, 0.5, 2.0, 5.0, 10.0 and 25.0μIU/mL Lyophilized, 1 set
  • TMB Reagent (One-Step), 1 vial (11mL)
  • Stop Solution (1N HCl): Natural cap, 1 vial (11mL)

The Abnova TSH ELISA test is based on the principle of a solid phase enzyme-linked immunosorbent assay

Specifications

Calibration Range 0 to 25uIU/mL
Conjugate HRP
Cross Reactivity Human
Description TSH (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of thyroid stimulating hormone.
Detection Limit 0.2uIU/mL
Detection Method Colorimetric
Includes Antibody-Coated Wells 96 wells, Enzyme Conjugate Reagent (13mL), Reference Standard Set: 0, 0.5, 2.0, 5.0, 10.0 and 25.0μIU/mL Lyophilized (1 set), TMB Reagent (11mL), Stop Solution (11mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Sensitivity 0.2 μIU/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.