Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TSH (Human) ELISA Kit

Catalog No. 89100209 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Serum
Calibration Range:
0 to 10uIU/mL
0 to 25uIU/mL
0.05 to 12μIU/mL
0.2 to 40μIU/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type Calibration Range
89-100-209 Serum 0.05 to 12μIU/mL
89-100-043 Serum 0 to 10uIU/mL
89-100-044 Serum 0 to 25uIU/mL
89-100-211 Serum 0.2 to 40μIU/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 89-100-209 Supplier Abnova™ Supplier No. KA0517
Add to Cart
Add to Cart

Includes

  • MICROTITER PLATE, 96 wells
  • CONJUGATE, 12mL
  • Standard, 12μIU/mL, 1 vial
  • Standard, 6μIU/mL, 1 vial
  • Standard, 2μIU/mL, 1 vial
  • Standard, 0.8μIU/mL, 1 vial
  • Standard, 0.2μIU/mL, 1 vial
  • Standard, 0μIU/mL, 1 vial
  • SUBSTRATE A Buffered solution with H2O2, 10mL
  • SUBSTRATE B Buffered solution with TMB, 10mL
  • STOP SOLUTION, 14mL

This ultra-sensitive TSH enzyme linked immunosorbent assay (ELISA) applies a technique called a quantitative sandwich immunoassay

Specifications

Calibration Range 0.05 to 12μIU/mL
Conjugate HRP
Cross Reactivity Human
Description TSH (Human) High sensitivity ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human TSH in serum.
Detection Limit 0.05μIU/mL
Detection Method Colorimetric
Includes Antibody coated microwells 96 wells, CONJUGATE (12mL), STANDARD (12, 6, 2, 0.8, 0.2, 0μIU/m for each vial), SUBSTRATE A (10mL), SUBSTRATE B (10mL), Stop Solution (14mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Sensitivity 0.05 μIU/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.