Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Urea Assay Kit
GSA_VA

Catalog No. 89028384 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-384 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-384 Supplier Abnova™ Supplier No. KA1652
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Reagent A, 12mL
  • Reagent B, 12mL
  • Urea standard, 1mL (50mg/dL)

For quantitative colorimetric urea determination in bronchoalveolar lavage fluid, cell culture, milk, plasma, serum, tissue culture, urine

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Calibration Range 0.08 to 100mg/dL
Content And Storage Store the kit at -20°C. For best results, store the components as described in the protocol. Avoid repeated freezing and thawing.
Description Urea Assay Kit is a quantitative colorimetric urea determination.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 12mL Reagent A, 12mL reagent B, 1mL 50mg/dL Urea standard
Quantity 1 Kit
Sample Type Bronchoalveolar Lavage Fluid, Cell Culture, Milk, Plasma, Serum, Tissue Culture, Urine
Sample Volume 5 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.