Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Urease Assay Kit

Catalog No. 89028355 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-355 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-355 Supplier Abnova™ Supplier No. KA1623
Add to Cart
Add to Cart

Quantitative determination of urease activity

  • Sample Type: Biological Sample
  • Sample Volume: 90μL

Functional Study

Specifications

Content And Storage Store the kit at 4°C.
Description Urease Assay Kit is a quantitative determination of urease activity.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 20mL assay buffer (pH 7), 12mL reagent A, 1.5mL Urea, 6mL reagent B, 100μL NH4Cl (50mM)
Quantity 1 Kit
Sample Type Biological Sample
Sample Volume 90 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.