Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Uric Acid Assay Kit

Catalog No. 89028383 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-383 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-383 Supplier Abnova™ Supplier No. KA1651
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Reagent A ,50mL
  • Reagent B, 6mL
  • Reagent C, 6mL
  • Standard: 10mg/dL, uric acid, 1mL
  • Blank Control, 1mL

For quantitative colorimetric uric acid determination at 590nm in plasma, serum, urine

Specifications

Calibration Range 0.22 to 30mg/dL
Content And Storage Store reagents at 4°C. Store standard and blank control at -20°C.
Description Uric Acid Assay Kit is a quantitative colorimetric uric acid determination at 590 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 50mL Reagent A, 6mL reagent B, 6mL reagent C, 1mL standard, 10mg/dL Uric Acid, 1mL blank control
Quantity 1 Kit
Sample Type Plasma, Serum, Urine
Sample Volume 20 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.