Learn More
Description
Includes
- WST-8 Reagent (powder) 1 vial/1 ea
- Electron Mediator Solution 1 vial/1.2mL
Specifications
Specifications
| Content And Storage | Store the kit at -20°C. |
| Description | WST-8 Cell Proliferation Assay Kit provides an easy to use tool for studying induction and inhibition of cell proliferation in any in vitro model. |
| For Use With (Application) | Functional Study |
| Includes | 1 vial WST-8 reagent (powder), 1.2mL electron mediator solution |
| Quantity | 1 Kit |
| Type | Cell Proliferation Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.