Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Xanthine Oxidase Assay Kit

Catalog No. 89024506 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-506 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-506 Supplier Abnova™ Supplier No. KA1386
Add to Cart
Add to Cart

Includes

  • XO Assay buffer (10X), 1 vial
  • XO Sample buffer (10X), 1 vial
  • Xanthine oxidase standard, 2 vials
  • XO Detector, 3 vials
  • XO Horseradish peroxidase, 2 vials
  • XO Dimethylsulfoxide, 1 vial
  • 96-Well plate (black), 1 plate
  • 96-Well cover sheet, 1 cover

Used for quantifying xanthine oxidase activity and functional study

Specifications

Content And Storage Store the kit at -20°C.
Description Xanthine Oxidase Assay Kit provides a simple and accurate method for quantifying xanthine oxidase activity.
For Use With (Application) Functional Study
Includes 1 vial XO assay buffer 10X, 1 vial XO sample buffer 10X, 2 vials Xanthine Oxidase standard, 3 vials XO Detector, 2 vials XO Horseradish Peroxidase, 1 vial XO Dimethylsulfoxide, 1 plate 96-well plate (black), 1 cover 96 well cover sheet
Quantity 1 Kit
Type Xanthine Oxidase Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.