Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ XTT Cell Proliferation Assay Kit

Catalog No. 89024507 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-507 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-507 Supplier Abnova™ Supplier No. KA1387
Add to Cart
Add to Cart

Includes

  • XTT Reagent (powder) 1 vial/1 ea
  • Electron Mediator Solution 1 vial/1.2mL

Used for studying induction and inhibition of cell proliferation in any in-vitro model and functional study

Specifications

Content And Storage Store the kit at -20°C.
Description XTT Cell Proliferation Assay Kit provides an easy to use tool for studying induction and inhibition of cell proliferation in any in vitro model.
For Use With (Application) Functional Study
Includes 1 vial XTT reagent (powder), 1.2mL electron mediator solution
Quantity 1 Kit
Type Cell Proliferation Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.