Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Zinc Assay Kit

Catalog No. 89028351 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Plasma (no EDTA), Saliva, Serum, Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-028-351 Plasma (no EDTA), Saliva, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-351 Supplier Abnova™ Supplier No. KA1619
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Reagent A, 50mL
  • Reagent B, 1mL
  • Reagent C, 1mL
  • EDTA, 1mL (100mM)
  • Zinc standard, 1mL (50μM)

For quantitative colorimetric zinc determination at 425nm

Specifications

Calibration Range 0.12 to 10μM
Content And Storage Store Reagents B and C at -20°C. Store other components at 4°C.
Description Zinc Assay Kit is a quantitative colorimetric zinc determination at 425 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 50mL Reagent A, 1mL reagent B, 1mL reagent C,1mL (100mM) EDTA, 1mL (50μM) Zinc standard
Quantity 1 Kit
Sample Type Plasma (no EDTA), Saliva, Serum, Urine
Sample Volume 50 μL
Type Zinc Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.