Learn More
Bullard™ AboveView™ Hard Hat with Replaceable Clear Brim Visor
SDP
Catalog No. 19160653 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Hi-vis Orange
Hi-vis Yellow
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-160-653 Hi-vis Orange
19-160-650 White
19-160-651 Yellow
19-160-654 Hi-vis Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-160-653 Supplier Bullard™ Supplier No. AVHORC
This item is not returnable. View return policy
For increased protection from the elements and harsh environments
  • Features see-through brim visor that allows workers to see hazards overhead
  • Designed with a wide brim for increased protection
  • 3/4 in. seamless suspension, large absorbent cotton or easy-to-clean cotton brow pad
  • Flex-Gear 4-point ratchet suspension, and front, side, and rear vertical height adjustment settings
  • Nylon crown straps
  • Cushioned nape
  • Accessory Slots
  • Available in standard colors and can be customized with striping and logos
Specifications
Color Hi-vis Orange
Includes Replaceable clear brim visor
Product Type Hard Hat
Suspension Type 4-point ratchet
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.