Learn More
R&D Systems™ AbPure™ Magnetic Beads Antibody Purification Kit
SDP
Catalog No. 2650200 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.4 mL
Catalog No. Quantity
26-502-00 0.4 mL
1 options
Catalog No. 26-502-00 Supplier R&D Systems™ Supplier No. 2650200
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
A Novus product, part of R&D Systems. R&D Systems™ AbPure Magnetic Antibody Purification Kit is designed to be compatible with Lightning-Link, Thunder-Link PLUS, InnovaCoat GOLD and LATEX conjugation kits.
Kit Components:
  • 400μL of Protein A Magnetic Beads
  • One vial of Neutralization Buffer
  • One bottle of Wash Buffer
  • One vial of 10X Binding Buffer
  • One vial of Elution Buffer
  • Not supplied magnetic stand
Specifications
Sufficient For One reaction
Product Type AbPure Magnetic Antibody Purification Kit
Content And Storage 400μL Protein A Magnetic Beads, 1 vial of neutralization buffer, 1 bottle of wash buffer, 1 vial of 10X binding buffer, 1 vial of elution buffer, Not supplied magnetic stand. 4°C
Product Line Novus
Quantity 0.4 mL
For research use only.