Learn More
Description
- Used to purify antibodies from crude samples such as ascites fluid or immune serum
- Antibody to be purified or cleaned up ideally is in volume of 0.1mL to 0.5mL
- 20 to 150μg of antibody can be purified in each run
Specifications
Specifications
| Sufficient For | 3 Purifications |
| Product Type | Antibody Purification Kit |
| Content And Storage | 1 or 3 vials AbPure Mouse resin, 1 or 3 spin cartridge/collecting tube assemblies, 1 vial 10x AbPure Binding buffer, 1 vial AbPure neutralization buffer, 1 vial AbPure wash buffer, 1 vial AbPure elution buffer, 4 or 12 additional collecting tubes. 4°C |
| Product Line | Novus |
| Quantity | 3 Tests |
For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
