Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Abpure™ TCS Antibody Purification Kit
SDP

Catalog No. 2640030 Shop All R&D Systems Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
26-400-30 1 Purification
26-405-00 3 Purifications
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-400-30 Supplier Novus Biologicals™ Supplier No. 2640030

Novus Biologicals™ Abpure TCS Antibody Purification Kit can be used in its rapid version by spin-purification in centrifuge, or by gravity-flow purification.

  • Fully compatible with InnovaCoat GOLD and LATEX conjugation kits which allow purified antibody to be conjugated rapidly
  • Antibody to be purified should be in 10mL to 50mL of tissue culture supernatant (TCS)
  • Up to 5mg of antibody can be purified in each run

Specifications

Content And Storage 4°C
Quantity 1 Test
Kit Contents 4 or 12 15mL collection tubes, 1 bottle of AbPure elution buffer, 1 bottle of AbPure neutralization buffer, 1 bottle of AbPure preservative, 1 bottle of 10x AbPure binding buffer, 1 bottle of AbPure wash buffer, 1 or 3 bottles of AbPure TCS protein A resi
Sufficient For 1 Purification
Product Type Antibody Purification Kit

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.