Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Abrin-a Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144392 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144392 Supplier Invitrogen™ Supplier No. PA5144392
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Abrin-a Polyclonal Antibody for Western Blot, ELISA

The A chain is responsible for inhibiting protein synthesis through the catalytic inactivation of 60S ribosomal subunits by removing adenine from position 4,324 of 28S rRNA. Abrin-a is more toxic than ricin.; The B chain is a galactose-specific lectin that facilitates the binding of abrin to the cell membrane that precedes endocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Abrin-a
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.432 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias Abrin-a
Host Species Rabbit
Immunogen Recombinant Abrus precatorius Abrin-a protein (1-251aa ).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.