Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abscisic acid (S)-(+) | 21293-29-8 | MFCD00066545 | 99.7% | 264.32 g/mol | C15H20O4 | 500 MG
SDP

Supplier:  Medchemexpress LLC HY100560500MG

Encompass_Preferred

Abscisic acid (S)-(+)- is a naturally occurring plant hormone supplied as a research-grade analytical standard. It is used to study plant growth regulation, signaling pathways, and stress responses, and is provided as a solid suitable for biochemical assays and metabolic studies.

  • Research-grade analytical standard for plant biology studies.
  • High purity: 99.7%.
  • Molecular formula C15H20O4; molecular weight 264.32 g/mol.
  • Supplied as a 500 MG solid for laboratory use.
  • Suitable for biochemical assays and metabolic studies.
  • Common applications: plant growth and stress-response research.

Catalog No. 50-002-12787


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.