Learn More
R&D Systems™ AbSelect™ Antibody Concentration and Clean Up Antibody Purification Kit
SDP
Catalog No. 8610010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
3 Tests
Catalog No. Quantity
86-100-10 3 Tests
1 options
Catalog No. 86-100-10 Supplier R&D Systems™ Supplier No. 8610010
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
A Novus product, part of R&D Systems. Allows quick and easy concentration of antibodies and proteins
The antibody clean up kit method utilizes a simple spin column to easily and quickly remove excess buffer from the antibody thereby providing a more concentrated antibody solution. The clean up kit also allows the experimenter to perform a simple buffer exchange to transfer the antibody into a more favorable conjugation buffer. The kit can also be used to reduce the concentration of many unwanted additives often found in antibody formulations such as azide, glycine or tris.
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Product Type Antibody Concentration and Clean-up Antibody Purification Kit
Product Line Novus
Content And Storage 4°C. Do not freeze.
Quantity 3 Tests