Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AbSelect™ G Antibody Purification Kit
SDP

Catalog No. p-6376305 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
89-000-05 1 Purification
89-000-10 3 Purifications
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-000-05 Supplier R&D Systems™ Supplier No. 8900005

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. For purification of antibodies from crude samples such as ascites fluid or immune serum and removal of contaminants

The method involves capturing the antibody on the AbSelect Protein G resin. Protein G has a high affinity for the Fc regions of IgG molecules from a variety of species and once the antibody has bound to the Protein G, unwanted substances can be removed by simply washing the resin. The purified product is then eluted and neutralized.

  •  Compatible with the lightning-link and lightning-link rapid conjugation systems
    • Allows purified antibody to be immediately labeled with a hands-on time of under 30 seconds

Recommended Storage

Store at 4°C. Do not freeze.

Specifications

Product Type Antibody Purification Kit
Product Line Novus
Sufficient For 1 Purification
Content And Storage 4°C. Do not freeze.
Quantity 1 Test
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.