Learn More
Description
Protein G has a high affinity for the Fc regions of IgG molecules from a variety of species. AbSelect G Serum resin is prepared by coupling purified Protein G to agarose beads. It can therefore be used to purify IgG fractions from both serum and ascites fluid.
- Compatible with the lightning-link antibody labeling kits
- Allows purified antibody to be immediately labeled with an enzyme, fluorescent protein or fluorescent dye, with a hands-on time of under 30 seconds
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Specifications
| Product Type | Antibody Purification Kit |
| Sufficient For | 3 Purifications |
| Content And Storage | 4°C. Do not freeze. |
| Quantity | 3 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
