Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AbSelect™ Generic Antibody Purification Kit
SDP

Catalog No. 8600010 Shop All Novus Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
86-000-10 3 Purifications
86-000-05 1 Purification
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 86-000-10 Supplier R&D Systems™ Supplier No. 8600010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Fast and simple method to purify antibodies from ascites fluid and serum

The AbSelect method involves capture of the antibody on protein A resin and the removal of unwanted substances by a simple wash procedure. The purified product is then eluted and neutralized. The antibody to be purified or cleaned up ideally is in a volume of 100μL to 0.5mL, up to 500μg of antibody can be purified in each run.

Compatible with lightning-link antibody labeling system, which allows purified antibody to be rapidly conjugated with wide variety of labels

Recommended Storage

Store at 4°C. Do not freeze.

Specifications

Product Type Antibody Purification Kit
Product Line Novus
Sufficient For 3 Purifications
Content And Storage 4°C. Do not freeze.
Quantity 3 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.