Learn More
Description
- Mouse antibody to be purified or cleaned up is in volume of 0.1 to 0.5mL
- Up to 150g of antibody can be purified in each run
- This system is not suitable for use with antibodies from other species
- Components of the AbSelect kit are fully compatible with lightning-link and lightning-link rapid antibody labeling system
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Specifications
| Product Type | Antibody Purification Kit |
| Product Line | Novus |
| Sufficient For | 3 Purifications |
| Content And Storage | 4°C. Do not freeze. |
| Quantity | 3 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
