Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AbSelect™ Mouse TCS Antibody Purification Kit
SDP

Catalog No. 8320005 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
83-200-05 1 Purification
83-205-00 3 Purifications
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 83-200-05 Supplier R&D Systems™ Supplier No. 8320005

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. For purification of mouse IgG fractions from hybridoma supernatants

AbSelect Mouse TCS resin has a high affinity for mouse IgG molecules. The method involves capture of the antibody on the AbSelect Mouse TCS resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.

  • This system is not suitable for use with antibodies from other species
  • Binding strength of Bovine IgG to AbSelect Mouse resin is negligible
  • Components of the AbSelect kit are fully compatible with lightning-link and lightning-link rapid antibody labeling system

Recommended Storage

Store at 4°C. Do not freeze.

Specifications

Product Type Antibody Purification Kit
Product Line Novus
Sufficient For 1 Purification
Content And Storage 4°C. Do not freeze.
Quantity 1 Test
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.