Learn More
Description
AbSelect Mouse TCS resin has a high affinity for mouse IgG molecules. The method involves capture of the antibody on the AbSelect Mouse TCS resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.
- This system is not suitable for use with antibodies from other species
- Binding strength of Bovine IgG to AbSelect Mouse resin is negligible
- Components of the AbSelect kit are fully compatible with lightning-link and lightning-link rapid antibody labeling system
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Specifications
| Product Type | Antibody Purification Kit |
| Product Line | Novus |
| Sufficient For | 1 Purification |
| Content And Storage | 4°C. Do not freeze. |
| Quantity | 1 Test |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
