Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AbSelect™ Serum Antibody Purification Kit
SDP

Catalog No. 8630030 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
86-300-30 1 Purification
86-305-00 3 Purifications
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 86-300-30 Supplier R&D Systems™ Supplier No. 8630030

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Fast and simple method to purify antibodies from ascites fluid and serum

AbSelect Serum is prepared by coupling highly purified protein A to agarose beads which can be used to purify IgG fractions from ascites fluid and serum. The method involves the capture of the antibody on the AbSelect Serum resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.

  •  Compatible with the lightning-link antibody labeling kits
  • Allows purified antibody to be immediately labeled with an enzyme, fluorescent protein or fluorescent dye

Recommended Storage

Store at 4°C. Do not freeze.

Specifications

Product Type Antibody Purification Kit
Product Line Novus
Sufficient For 1 Purification
Content And Storage 4°C. Do not freeze.
Quantity 1 Test
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.