Learn More
Description
AbSelect TCS is prepared by coupling highly purified protein A to agarose beads which can be used to purify IgG fractions from hybridoma supernatants. The method involves the capture of the antibody on the AbSelect TCS resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.
- Purify antibody from 10 to 50mL of hybridoma TCS
- 100% compatible with lightning-link range of conjugation kits
- Supplied with all components and buffers
- Available with full technical support
Recommended Storage
Store at 4°C. Do not freeze
Specifications
Specifications
| Product Type | Antibody Purification Kit |
| Product Line | Novus |
| For Use With (Application) | The antibody to be purified should be in 10 to 50 mL of tissue culture supernatant. Up to 5 mg of antibody can be purified in each run. |
| Sufficient For | 1 Purification |
| Content And Storage | 4°C. Do not freeze. |
| Quantity | 1 Test |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
