Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Absolute Q™ Liquid Biopsy dPCR Assay - EGFR 6253

Catalog No. A52797
Encompass_Preferred
Change view
Click to view available options
Absolute Q Liquid Biopsy dPCR Assay - EGFR 6253, 700 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A52797 Absolute Q Liquid Biopsy dPCR Assay - EGFR 6253, 700 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A52797 Supplier Applied Biosystems™ Supplier No. A52797
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Absolute Q™ Liquid Biopsy dPCR assays enable reproducible, specific detection of known somatic mutations.

Each predesigned Absolute Q™ Liquid Biopsy dPCR assay has been validated using controls to detect down to 0.1% variant allele frequency in genes relevant for cancer research.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 40X
Detection Method Digital PCR
Format Tube
PCR Method Digital PCR
Product Line Absolute Q
Compatibility QuantStudio™ Absolute Q™ Digital PCR System
Final Product Type Assay
No. of Reactions 700
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.