Learn More
New Pig Absorbent Mat Roll
SDP
Catalog No. 18389132 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Absorbent Mat Roll
Catalog No.
18-389-132 Type: Absorbent Mat Roll
1 options
This item is not returnable. View return policy
Catalog No. 18-389-132 Supplier New Pig Supplier No. MAT230
  • $318.00
    $225.65 / Each of 1
    Save $92.35
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
This mat lasts 2x longer than ordinary mats for fewer change-outs. It absorbs oils, coolants, solvents and water; just about any mess you need to clean up!
  • Lasts 2X longer than ordinary mats for fewer change-outs; that's why more plants worldwide use PIG Mat than any other brand
  • Multiple layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind
  • Absorbs most common industrial liquids - oils, water, solvents, coolants and more
  • Absorbed leaks and drips blend right in; dark gray color hides grime so mat stays on the job longer
  • Can be wrung out and incinerated after use to reduce waste or for fuels blending
  • Rolls are ideal for covering large areas and soaking up leaks and overspray
  • Easy-tear perforations let you take only what you need, so you use less mat and save money