Learn More
PIG™ Absorbent Pillow
SDP
Catalog No. 18808757 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Cellulose, Polypropylene
Catalog No. Material
18-808-757 Cellulose, Polypropylene
1 options
Catalog No. 18-808-757 Supplier PIG™ Supplier No. PIL204
Add to Cart
Add to Cart
When the situation calls for more than a mat, our high-capacity pillows deliver the absorbency you need.
  • Pillow has large surface area, high capacity and fast-wicking filler to quickly soak up liquids
  • Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated
  • Cellulose filler is highly absorbent for long-term use or big spill cleanup
  • Compact design is great for catching persistent drips and leaks in small spaces or adding absorbency for spill response
  • Absorbs most common industrial liquids - oils, water, solvents, coolants; not recommended for acids, bases or other corrosive liquids
  • Recycled content is ideal when green products are desired or required
  • Can be incinerated after use to reduce waste or for fuels blending
Specifications
Material Cellulose, Polypropylene
Disposable Single-use
Weight Capacity Heavyweight