Learn More
Description
Includes
A filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).
Fit standard spectrophotometer cell holders
- All cells are 22mm wide
- Available size is macro
- 10, 20, 50 or 100mm path length rectangular cells
- Available with a variety of window material for wide range of assays
- Available in two materials e.g. Quartz and Special Optical Glass
- Quartz windows for transmission from far ultraviolet through the infrared
- Glass windows for routine work in ultraviolet region and visible region
Specifications
Specifications
| Type | Cell |
| Capacity (Metric) | 6300 μL |
| Material | Quartz |
| Width (Metric) Interior | 18.5mm |
| Path Length | 20 mm |
| Wavelength Range | 190 to 2600 nm, 2850 to 3600 nm |
| Height (Metric) | 35 mm |
| Width (Metric) | 22 mm |
| Shape | Cylinder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.