Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Absorption Cylindrical Cells

Catalog No. 14958142 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
16,000 μL
31,000 μL
3100 μL
6300 μL
Material:
Optical Glass
Quartz
Special Optical Glass
Wavelength Range:
190 to 2600 nm, 2850 to 3600 nm
320 to 2500 nm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Material Wavelength Range
14-958-142 16,000 μL Quartz 190 to 2600 nm, 2850 to 3600 nm
14-958-147 31,000 μL Optical Glass 320 to 2500 nm
14-958-140 3100 μL Quartz 190 to 2600 nm, 2850 to 3600 nm
14-958-146 16,000 μL Special Optical Glass 320 to 2500 nm
14-958-143 31,000 μL Quartz 190 to 2600 nm, 2850 to 3600 nm
14-958-145 6300 μL Optical Glass 320 to 2500 nm
14-958-141 6300 μL Quartz 190 to 2600 nm, 2850 to 3600 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 14-958-142
This item is not returnable. View return policy

Includes

A filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).

Fit standard spectrophotometer cell holders

  • All cells are 22mm wide
  • Available size is macro
  • 10, 20, 50 or 100mm path length rectangular cells
  • Available with a variety of window material for wide range of assays
  • Available in two materials e.g. Quartz and Special Optical Glass
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Specifications

Type Cell
Capacity (Metric) 16,000 μL
Material Quartz
Width (Metric) Interior 18.5mm
Path Length 50 mm
Wavelength Range 190 to 2600 nm, 2850 to 3600 nm
Height (Metric) 35 mm
Width (Metric) 22 mm
Shape Cylinder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.