Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ AbSurance™ Complete Core Histone Antibody Specificity Array
Shop All MilliporeSigma ProductsDescription
Requires
Primary Antibody, Secondary Antibody, Methanol, TBST Solution, Blocking Solution (5 % non-fat dried milk in TBST), Immobilon™ Western Chemiluminescent HRP Substrate, or RapidStep™ ECL Reagent, Containers for the membrane, Shaker platform, Micropipettes and pipette tips, Film or Imager
- Simple Western blot-like detection procedure
- 〉95 % pure peptides analyzed by mass spectrometry
- Two concentrations of peptides allows detection of both strong and weak cross-reactivity
- Consistent and uniform spotting of peptides using proprietary process
- Data collection using either film or standard CCD imaging systems
- Easy- to-read 8 x 12 macroarray
- PVDF-FL membrane allows sensitive chemiluminescent or fluorescent detection
- Positive control IgG spots and membrane landmarks for simple and easy spot identification
Specifications
Specifications
| Content And Storage | Room Temperature up to 6 Months |
| Detection Method | Chemiluminescence, Fluorescence |
| For Use With (Application) | Antibody specificity screening of histone peptides |
| Quantity | 2 Arrays |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?