Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ABT 263
GREENER_CHOICE

Catalog No. 768010
Change view
Click to view available options
Quantity:
10 mg
50 mg
Catalog No. Quantity
76-801-0 10 mg
76-805-0 50 mg
2 options
This item is not returnable. View return policy

Catalog No. 76-801-0

Supplier: Tocris Bioscience™ 7680/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity Bcl-2 family inhibitor; proapoptotic, senolytic and antitumor

ABT 263 is a high affinity inhibitor of Bcl-2 family proteins (Ki values are 0.5 nM, <1 nM and <1 nM in Bcl-xL, Bcl-2 and Bcl-W respectively). ABT 263 induces G1/G0 phase arrest, apoptosis and autophagy in cancer cells in vitro (IC50 = 3.4 - 12 μM). In vivo, ABT 263 displays a synergistic antitumor effect with rituximab and Bortezomib (Cat. No. 7282), Docetaxel (Cat. No. 4056) and Erlotinib (Cat. No. 7194), and inhibits growth of SCLC xenografts when combined with (+)-JQ1 (Cat. No. 4499). ABT 263 also reduces inflammation and viral-induced senescence in SARS-CoV-2 infection in vivo, and clears senescent cells and rejuvenates aging stem cells. Orally bioavailable.

Specifications

Chemical Name or Material 4-[4-[[2-(4-Chlorophenyl)-5,5-dimethyl-1-cyclohexen-1-yl]methyl]-1-piperazinyl]-N-[[4-[[(1 R)-3-(4-morpholinyl)-1-[(phenylthio)methyl]propyl]amino]-3-[(trifluoromethyl)sulfonyl]phenyl]sulfonyl]benzamide
CAS 923564-51-6
Quantity 10 mg
Target Bcl-2 Family Inhibitors
Molecular Formula C47H55ClF3N5O6S3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.