Learn More
Medchemexpress LLC Abt-418 hydrochloride | 147388-83-8 | MFCD11045960 | 99.4% | 202.68 g/mol | C9H15ClN2O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY105170B50MG
Encompass_Preferred
ABT-418 hydrochloride is a potent, selective nicotinic acetylcholine receptor (nAChR) agonist supplied as the hydrochloride salt for preclinical research. It exhibits cognitive-enhancing and anxiolytic activities and is used in pharmacology and receptor studies.
Catalog No. 50-002-43606
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.