Learn More
TARGETMOL CHEMICALS INC Sonlicromanol hydrochloride
Supplier: TARGETMOL CHEMICALS INC T380131MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Sonlicromanol hydrochloride (KH176 hydrochloride) is a ROS-redox modulator that crosses the blood-brain barrier, has potential anticancer activity, improves m.3243A>G heterogeneity in iPSC-derived neurons associated with It has potential anticancer activity, ameliorates neuronal network dysfunction and transcriptomic changes associated with m.3243A>G heterogeneity in iPSC-derived neurons, and can be used for the treatment of mitochondrial diseases. Purity 99.53%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.