Learn More
Eurofins Discoverx ABT-737, 5 mg. (92-1151)
Supplier:  Eurofins Discoverx 921151
Encompass_Preferred
ABT-737 is a small molecule BH3 mimetic inhibitor of the anti-apoptotic members of the BCL-2 family, selective for BCL-2, BCL-XL, and BCL-W (EC50=78.7, 30.3, and 197.8 nM, respectively). Developed by AbbVie, it failed in clinical trials due to toxicity issues, but was proven efficacious with various radiation therapies in triggering apoptosis in cancer cells and xenografts. Provided as 5 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9682
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.