Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABTS 98.0+%, TCI America™
SDP

Catalog No. A21661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21661G Supplier TCI America Supplier No. A21661G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[for Biochemical Research]

Specifications

CAS 30931-67-0
Color Green-White
Molecular Formula C18H24N6O6S4
Quantity 1 g
MDL Number MFCD00010404,MFCD00010404
Synonym ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
InChI Key OHDRQQURAXLVGJ-AXMZSLBLSA-N
SMILES [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O
IUPAC Name diammonium (2Z)-3-ethyl-2-{2-[(2Z)-3-ethyl-6-sulfonato-2,3-dihydro-1,3-benzothiazol-2-ylidene]hydrazin-1-ylidene}-2,3-dihydro-1,3-benzothiazole-6-sulfonate
Molecular Weight (g/mol) 548.67
PubChem CID 91884754
Formula Weight 548.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material ABTS [=2,2′-Azinobis(3-ethylbenzothiazoline-6-sulfonic Acid Ammonium Salt)] [for Bioche
Show More Show Less
RTECSNumber DL7002000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.