Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ABTS™ Chromophore, Calbiochem™,

Catalog No. 194434100ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
19-443-4100ML 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-443-4100ML Supplier MilliporeSigma™ Supplier No. 194434100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C18H18N4O6S4
Synonym 2,2'-Azino-bis(3-ethylbenzthiazoline-6-sulfonic Acid)

Specifications

Color Yellow
Packaging Glass Bottle
Quantity 100 mL
Physical Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.