Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABTS ELISA Buffer Kit, PeproTech®
Description
- Contains the key components required for the analysis of ten sandwich ELISA plates
- Specifically formulated to be used in conjunction with PeproTech standard ELISA development kits
Included solutions and materials:
- PBS
- Wash buffer
- Blocking buffer
- Diluent
- ABTS liquid substrate
- ELISA microplates
- Sealing film
Specifications
Specifications
| Product Type | ELISA Buffer |
| Quantity | 1 Kit |
| Detection Method | Colorimetric |
| For Use With (Application) | ELISA |
| Product Line | PeproTech |
| Substrate Type | ABTS, HRP |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?