Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AC 265347
GREENER_CHOICE

Catalog No. p-7114666
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-651-0 10 mg
61-655-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-651-0 Supplier Tocris Bioscience™ Supplier No. 6165/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

CaSR biased allosteric modulator

AC 265347 is a CaSR biased allosteric modulator (pEC50 = 7.8-8.1). Calcimimetic. Displays ability to bias signalling towards the accumulation of pERK1/2 and IP1. Reduces parathyroid hormone (PTH) levels in rat serum without inducing the release of calcitonin. Exhibits no significant activation of human GABAB or type I PTH receptors. Orally active.

Specifications

Chemical Name or Material α-(2,4-Dimethylphenyl)-α-methyl-2-benzothiazolemethanol
CAS 1253901-26-6
Assay Percent Range >99%
Quantity 10 mg
Molecular Formula C17H17NOS
Formula Weight Observed MW: 283.39
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.