Learn More
Description
- Hydrolysis of Lys-AMC amide bond releases AMC, a highly fluorescent group
- A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
- After the enzymatic removal of the acetyl group, the substrate can be cleaved by trypsin-like enzymes
- Purity >95%
Specifications
Specifications
| Product Line | R&D Systems |
| Quantity | 5 mg |
| Product Type | Ac-Arg-Gly-Lys(Ac)-AMC (Ac-RGK(Ac)-AMC) |
| Storage Requirements | Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
