Learn More
CPC Scientific Ac-Asp-Glu-Val-Asp-AFC 5MG
Supplier: CPC Scientific CASP048B
Sensitive fluorescent substrate for the assay of caspase-3 activity. When halogen substituents are present, membrane permeability rises.ONE-LETTER SEQUENCE: Ac-DEVD-AFCMOLECULAR FORMULA: C30H34F3N5O13MOLECULAR WEIGHT:729.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [201608-14-2], RESEARCH AREA: ApoptosisREFERENCES: J. Xiang et al., Proc. Natl. Acad. Sci. USA, 93, 14559 (1996); H.Ohnimus, Personal communication (1997); J. Sun et al., Biochem. Biophys. Res. Commun., 238, 920 (1997)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.