Learn More
CPC Scientific Ac-Asp-Met-Gln-Asp-AMC 5MG
SDP
Supplier:  CPC Scientific CASP053B
Encompass
ONE-LETTER SEQUENCE: Ac-DMQD-AMCMOLECULAR FORMULA: C30H38N6O12S1MOLECULAR WEIGHT:706.73STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [355137-38-1], SYNONYMS: Caspase 3 Substrate 1m, fluorogenicRESEARCH AREA: Apoptosis
Catalog No. 50-210-9013
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.